Autosampler Syringes for Shimadzu AOC 14, 17, 20, and 20i GCs, SGE, Restek
Supplier: Restek
24844 24845
10059-306EA 311.09 CAD
10059-306 10059-308
Autosampler Syringes for Shimadzu AOC 14, 17, 20, and 20i GCs, SGE, Restek
Syringes Syringes for Chromatography
SGE syringes are designed and tested to meet critical autosampler performance needs.
  • Compatible with Shimadzu AOC 14, 17, 20, and 20i GCs
Needle point styles are designed to withstand multiple rapid injections through a septum.