Cylinder Holders, Wall Mounted, Restek
Supplier: Restek
21333 23400 23401 23402
10855-760EA 189.3 CAD
10855-760 10855-764 10855-766 10855-768
Cylinder Holders, Wall Mounted, Restek
Wall Hooks
These holders are designed to prevent free-standing gas cylinders from tipping over and injuring personnel.
  • Cylinder holders can accommodate 1 to 4 cylinders depending on model
The cast aluminum holder can be secured to a wall or the side of a work bench. Each mount will secure a cylinder 4 to 15 inches in diameter.