Graphite Sealing Ring for Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL instruments; fits 8 mm OD liners, Restek
Supplier: Restek
21899 21898
RK21899PK 97.24 CAD
RK21899 10852-576
Graphite Sealing Ring for Thermo Scientific TRACE, 8000, 8000 TOP & Focus SSL instruments; fits 8 mm OD liners, Restek
Seals Mechanical Seals
Graphite sealing rings are ideal for use with Thermo Scientific TRACE, 8000, 8000 TOP, and Focus SSL instruments.
  • Compatible with 8 mm OD liners
  • Durable graphite
  • Max operating temperature of 450°C