Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
20883 21884 21878 21879
RK20883PK 93.3 CAD
RK20883 RK21884 10803-260 10855-916
Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Fittings and Connectors
Capillary column nuts are available in brass or stainless steel.
  • Designed to fit a wider variety of 1/16" ferrules
For use with Agilent 5890/6850/6890/7890 GCs.