Capillary Dedicated FID Replacement Jet for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
21683 23079 21621 21682 23078 21620
RK21683PK 523.59 CAD
RK21683 10058-856 10803-250 10803-252 10803-310 10855-908
Capillary Dedicated FID Replacement Jet for Agilent 5890/6850/6890/7890 GCs, Restek
Chromatography Supplies Chromatography Spare Parts
Available in standard or Siltek®-treated high-performance versions.
  • Threads specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • Meets or exceeds original manufacturer's performance
For use with Agilent 5890/6850/6890 GCs. Can be Siltek® treated to be extremely inert, for use with active compounds.