Capillary Adaptable FID Replacement Jet for Agilent 5890/6850/6890 GCs, Restek
Supplier: Restek
20671 20672 20670 20673
RK20671PK 508.72 CAD
RK20671 RK20672 RK20670 RK20673
Capillary Adaptable FID Replacement Jet for Agilent 5890/6850/6890 GCs, Restek
Chromatography Supplies Chromatography Spare Parts
Available in standard or Siltek®-treated high-performance versions.
  • Threads specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • Meets or exceeds original manufacturer's performance
For use with Agilent 5890/6850/6890 GCs. Can be Siltek® treated to be extremely inert, for use with active compounds.