Septum Nuts for use with Split/Splitless Injectors, Restek
Supplier: Restek
21309 20631
RK21309EA 133.52 CAD
RK21309 RK20631
Septum Nuts for use with Split/Splitless Injectors, Restek
Fittings and Connectors
Septum nuts are for use with Agilent 5890/6850/6890/7890 split/splitless injectors.
  • Needle guide allows easy penetration and prevents septum coring
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections
  • Made of high-quality stainless steel