Cross-Disk Inlet Seals for Agilent GCs, Restek
Supplier: Restek
21012 20476 20477 20474 20475 21009 21010 21011
RK21012PK 718.87 CAD
RK21012 RK20476 10058-560 10803-218 10855-854 10855-868 10855-870 10855-872
Cross-Disk Inlet Seals for Agilent GCs, Restek
Seals Mechanical Seals
Cross-disk inlet seals are treated to provide enhanced inertness for use in a variety of applications.
  • Ideal for high-flow split applications >500 mL/min on Agilent 5890 GCs
  • Siltek® treatment provides enhanced inertness versus stainless steel
  • All seals include washers
Ideal for high-flow split applications, these inlet seals are gold-plated or Siltek®-treated.