Split/Splitless Injection Port for Agilent 5890 GCs, Direct Replacement or Siltek® Treated, Restek
Supplier: Restek
20266 20268
RK20266EA 666.53 CAD
RK20266 10855-850
Split/Splitless Injection Port for Agilent 5890 GCs, Direct Replacement or Siltek® Treated, Restek
Chromatography Supplies Chromatography Spare Parts
Siltek®-treated split/splitless injector is a direct replacement for Agilent 5890 and 6850/6890 GCs, but Siltek® treatment passivates the metal surface to ensure an inert pathway for the sample, delivering increased performance.
For use with manual flow or EPC on Agilent 5890 GCs.