Zero-Dead-Volume Valco® Internal Reducing Union, Restek
Supplier: Restek
20151 20152 20153
10852-910EA 135.11 CAD
10852-910 10852-912 10852-914
Zero-Dead-Volume Valco® Internal Reducing Union, Restek
Fittings and Connectors
Connect two sizes of tubing, using zero-dead-volume fittings on each end.
  • For 1/8" to 1/16" OD tubing
  • Stainless steel ferrules included
Designed so that no volume is introduced by the fitting which is not cleanly swept.