Zero Dead Volume Valco® Internal Union, Restek
Supplier: Restek
20148 20150 20147 20149
10059-060EA 152.49 CAD
10059-060 10059-062 10852-946 10852-948
Zero Dead Volume Valco® Internal Union, Restek
Fittings and Connectors
Valco® connectors allow for internal union within zero dead volume connectors.
  • Ends of tubing seat squarely at bottoms of fitting details
  • Made of 300-series stainless steel
  • For use with 1/16" OD tubing
  • Stainless steel ferrules included
Designed so that no volume is introduced by the fitting which is not cleanly swept.