Valco® Adapter Ferrules, ¹/₃₂", Valcon Polyimide, Restek
Supplier: Restek
20141 20140 20137
10059-058PK 231.85 CAD
10059-058 10803-208 10852-926
Valco® Adapter Ferrules, ¹/₃₂", Valcon Polyimide, Restek
Fittings and Connectors
Fused silica adaptor ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C.
  • Durable
  • Fabricated from Valcon polyimide
The determining factor for selecting adapter ferrule size is the fused silica tubing OD.