MXT® Low Dead Volume 'Y' Connector Kits for Metal Columns, Restek
Supplier: Restek
20396 20395 20537
RK20396EA 361.8 CAD
RK20396 10852-982 10852-984
MXT® Low Dead Volume 'Y' Connector Kits for Metal Columns, Restek
Fittings and Connectors
MXT® low dead volume Y connector kits for metal columns can be used to connect a guard column or transfer line to an MXT® stainless steel column.
  • Connect a guard column/transfer line to an MXT® stainless steel column
  • Low thermal mass tracks rapid oven temperature programming
  • Stainless steel ferrules and nuts
  • Siltek® treatment ensures ultimate inertness
Connect two MXT® columns to one inlet or one MXT® column to two detectors. Each kit contains the MXT® union, three 1/32-inch ferrules and nuts.