Universal Press-Tight® Connectors, Restek
Supplier: Restek
20401 20430 20400 20429 20480 20449 20402
RK20401PK 451.8 CAD
RK20401 RK20430 RK20400 RK20429 10803-220 10852-952 10852-980
Universal Press-Tight® Connectors, Restek
Fittings and Connectors
Press-Tight® connectors can be used to make multiple connections between columns and transfer lines, or to repair damaged columns.
  • Connect a guard column to an analytical column
  • Repair a broken column
  • Connect a column outlet to a transfer line
  • Deactivated fused quartz helps eliminate activity for complete inertness along the GC flow path
  • Universal fit: use with 0.1–0.53 mm ID Restek columns (0.33–0.74 mm OD)
Optional Siltek® deactivation is ideal for organochlorine pesticides analysis.