9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
Supplier: Restek
24485 24486 24498 24474 24473 24487 24497 24503 24505 24494 24504 24668 24488 24493 24495 24496 24500 24501 24502 24506 26598 24669 24670 26572 26573 26574 26575 26576 26577 26578 26580 26581 26599
RK24485PK 74.59 CAD
RK24485 RK24486 RK24498 RK24474 10059-164 10059-166 10059-168 10059-170 10059-172 10803-356 10803-358 10853-892 10853-956 10853-958 10853-960 10853-962 10853-964 10853-966 10853-968 10853-970 10854-262 10854-264 10854-266 10854-306 10854-308 10854-310 10854-312 10854-314 10854-316 10854-318 10854-320 10854-322 10854-384
9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
Closures Chromatography Vial Closures
9 mm short-cap closures are compatible with screw-thread or ribbed designs.
  • Economical
  • Sturdy polypropylene construction
These screw caps are for use with vials featuring short external threads where standard-length caps would be too long.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Order Now
SPECIFICATIONS
Environmentally Preferable