Reducing Ferrules, Restek
Supplier: Restek
20257 20217 20204 20203
RK20257PK 58.84 CAD
RK20257 RK20217 RK20204 10852-582
Reducing Ferrules, Restek
Fittings and Connectors
Reducing ferrules have a smoother surface and cleaner edges than conventional graphite ferrules.
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite or Vespel®/graphite material
  • Stable to 400°C
Available in graphite or Vespel®/graphite material.