WISP 48 Caps and Septa, Restek
Supplier: Restek
21743 21741 21749 21751 21742 21744 21745 21746 21750 21752 26588 26589
RK21743PK 103.43 CAD
RK21743 10058-680 10058-682 10058-684 10854-058 10854-060 10854-062 10854-064 10854-066 10854-068 10854-258 10854-260
WISP 48 Caps and Septa, Restek
Closures Chromatography Vial Closures
These replacement caps and septa are compatible with WISP vials (largely screw-thread) and serve as economical alternatives to purchasing new whole units.
  • Economical
  • Sturdy construction
  • Wide orifices for easier needle insertion
Durable, functional features make these inexpensive replacement parts an indispensable addition to any set of WISP vials.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.