WISP 48 Screw-Thread Step Vials, Restek
Supplier: Restek
24654 24656 24655 24657
RK24654PK 116.16 CAD
RK24654 10059-240 10853-882 10853-886
WISP 48 Screw-Thread Step Vials, Restek
Vials
WISP 48 screw-thread step vials feature a screw-thread design and an extra-wide orifice size of 13 mm.
  • Feature white graduated markings
  • Clear or amber color
  • Screw-thread format
Vials have a 4 ml capacity.