GRAPHPACK® 3D/2 Ferrules, Restek
Supplier: Restek
20271 20274
10803-212PK 686.29 CAD
10803-212 10852-968 10852-976
GRAPHPACK® 3D/2 Ferrules, Restek
Fittings and Connectors
Gerstel GRAPHPACK® technology provides a complete system that quickly and reliable makes leak-tight, low-dead-volume connections.
  • Ferrule internal diameter of 0.4 mm
Metal-jacketed graphite ferrules are the ideal seal for GC applications.