8 mm Autosampler Vial Convenience Kits, Restek
Supplier: Restek
21192 21194 24641 24638 24639 21193 21195
RK21192PK 141.58 CAD
RK21192 RK21194 10803-362 10853-910 10853-912 10854-036 10854-038
8 mm Autosampler Vial Convenience Kits, Restek
Vials
Autosampler vial kits are compartmentalized for convenient separation of parts for easy assembly.
  • Clear or amber glass vials
  • Treated or untreated
  • Black caps, red PTFE/silicone septa
8 mm vials have 2 ml capacity.