Caps and Septa, 8 mm, Preassembled, Restek
Supplier: Restek
21149 21720 21721 21722 26582 26583 26584 26585
10058-618PK 80.73 CAD
10058-618 10803-254 10854-042 10854-044 10854-246 10854-248 10854-250 10854-252
Caps and Septa, 8 mm, Preassembled, Restek
Closures Chromatography Vial Closures
Preassembled Restek screw caps with septa are available for creating dependable seals on vials with 8-millimeter-wide openings.
  • Screw-thread design
  • Economical
  • Black color caps
Black screw-thread caps are available to fit all 2.0 mL, 8 mm vials.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.