Mininert® Precision Sampling Valves for Vials, Restek
Supplier: Restek
24903 24904 24905 24901 24902
10853-926PK 310.67 CAD
10853-926 10853-928 10853-930 10854-292 10854-294
Mininert® Precision Sampling Valves for Vials, Restek
Valves Injection Valves
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum.
  • Screw-thread and crimp-top vial compatibility
  • Variety of orifice sizes
Models are available for screw-cap and crimp-top vials. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial. Turn the threaded flange to secure a tight fit.