GC Snap Ring Cap With Septum, 11 mm, Restek
Supplier: Restek
21731 21733 21734 21729 21730 21732
10058-674PK 86.46 CAD
10058-674 10058-676 10058-678 10854-052 10854-054 10854-056
GC Snap Ring Cap With Septum, 11 mm, Restek
Closures Chromatography Vial Closures
Restek snap ring caps are an extremely sturdy, affordable option when crimp-top caps are not needed.
  • Economical
  • Sturdy polypropylene construction
Use these snap caps when you crimping is unnecessary. Snap caps are available with either liners or septa, and they seal an 11-millimeter opening.