Aluminum Chloride Anhydrous

Supplier: ALDON CORP SE
AA0166-100GR
470300-122EA 85.8 CAD
470300-122
Aluminum Chloride Anhydrous
Chemicals for Science Education
CAS Number: 7446-70-0
Formula Weight: 133.34
Formula: AlCl3
Density (g/mL): 2.44
Boiling Point (°C): 182
Freezing Point (°C): 194
Solubility: Water
Shelf Life (months): 12
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR