Carborundum

Supplier: ALDON CORP SE

CC0184-100G CC0185-100G
470300-660EA 40.7 CAD
470300-660 470300-662
Carborundum
Chemicals for Science Education

CAS Number: 409-21-2
Formula Weight: 40.10
Formula: SiC
Density (g/mL): 3.2
Solubility: Insoluble in Water
Synonyms: Silicon Carbide
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR