SP Bel-Art No-Wire™ Round Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
18743-1016 F18743-1020
60915-544CS 368.89 CAD
60915-544 60915-548
SP Bel-Art No-Wire™ Round Racks, Bel-Art Products, a part of SP
Racks Tube Racks
The No-Wire™ racks fit neatly into round containers such as ice buckets and beakers, as well as desiccators.
  • Rack remains stable at the bottom of water baths or ice baths
  • Special polypropylene blend construction
  • All racks have a base and two tiers of grids
  • Numbered and lettered rows for identification
  • Easy to stack
  • Removable, snap-on loop handle for easy retrieval
  • Steam autoclavable at 121 °C (250 °F)
  • Racks are shipped flat and can be easily assembled
Choose from two sizes; the large white rack is ideal for use in round ice buckets such as the Magic Touch™, and the small red racks fit perfectly in a 1000 mL beaker. The top deck height for all racks is 7 cm (2.75").