Magnesium Nitrate 6-Hydrate

Supplier: ALDON CORP SE

MM0090-CAPS MM0090-100G MM0090-500G MM0090-100GR MM0096-500ML MM0090-PK/4R
470301-660EA 20.95 CAD
470301-660 470301-662 470301-664 470301-666 470301-670 470301-672
Magnesium Nitrate 6-Hydrate
Chemicals for Science Education

CAS Number: 13446-18-9
Formula Weight: 256.41
Formula: Mg(NO3)2·6H2O
Hazard Info: Oxidizer, Flammable, Irritant
Density (g/mL): 1.46
Boiling Point (°C): Decomposes
Freezing Point (°C): 89
Solubility: Water and Alcohol
Synonyms: Magnesium Nitrate Hexahydrate
Shelf Life (months): 36
Storage: Yellow


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR