Copper

Supplier: ALDON CORP SE

CC0420-500G CC0415-500G CC0414-100G CC0416-100G CC0416-500G CC0422-500GR CC0417-100G CC0417-500G CC0417-500GR CC0413-100G CC0413-500G CC0418-1SH CC0419-1SH CC0420-100G CC0425-100G CC0425-500G
470300-926EA 97.35 CAD
470300-926 470300-900 470300-902 470300-904 470300-906 470300-908 470300-910 470300-912 470300-914 470300-916 470300-918 470300-920 470300-922 470300-924 470300-928 470300-930
Copper
Chemicals for Science Education

CAS Number: 7440-50-8
Formula: Cu
Density: 8.92 g/mL
Boiling Point: 2595°C
Freezing Point: 1083°C
Synonyms: Copper Metal
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR