Cyclohexane

Supplier: ALDON CORP SE

CC0580-500MLR CC0580-1L CC0580-3.8L
470300-954EA 44.65 CAD
470300-954 470300-950 470300-952
Cyclohexane
Chemicals for Science Education

CAS Number: 110-82-7
Formula Weight: 84.16
Formula: CH2(CH2)4CH2
Hazard Info: Flammable, Toxic
Density (g/mL): 0.784
Boiling Point (°C): 82
Freezing Point (°C): 7
Solubility: Organic Solvents
Synonyms: Hexahydrobenzene
Shelf Life (months): 12
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR