Glycerin

Supplier: ALDON CORP SE

GG0120-1L GG0120-100ML GG0120-500ML GG0120-500MLR GG0120-50ML GG0120-3.8L
470301-162EA 66.75 CAD
470301-162 470301-164 470301-170 470301-174 470301-160 470301-168
Glycerin
Chemicals for Science Education

CAS Number: 56-81-5
Formula Weight: 92.10
Formula: HOCH2CHOHCH2OH
Hazard Info: Allergen
Density (g/mL): 1.2607
Boiling Point (°C): 290
Freezing Point (°C): 18
Solubility: Water and Alcohol
Synonyms: Glycerol
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR