Starch

Supplier: ALDON CORP SE

SS0960-500G SS0940-500GR SS0940-100GR SS0982-250ML SS0945-500ML SS0940-CAPS SS0940-CAP SS0940-100G SS0940-POUCH
470302-752EA 66.3 CAD
470302-752 470302-754 470302-760 470302-766 470302-768 470302-746 470302-748 470302-756 470302-770
Starch
Chemicals for Science Education

CAS Number: 9005-84-9 soluble, 9005-25-8 potato
Formula Weight: 162.15n
Formula: C6H10O5n
Density: 1.45 g/mL
Synonyms: Amylodextrin, Amylogen
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR