Support Stand and Clamps for Sonifier® Cell Disruptors, Branson Ultrasonics
Supplier: Branson Ultrasonics
101-063-1108 101-063-1109 101-063-1110
75795-832EA 135.45 CAD
75795-832 75795-834 75795-836
Support Stand and Clamps for Sonifier® Cell Disruptors, Branson Ultrasonics
Retort Stands
The support stand with stainless steel rod accommodates Branson’s Sonifier® line and accessories
  • Stand base measures 22.9L x 13.9W x 60.9H cm (9 x 5.5 x 24")
  • Stainless steel rod measures 60.9H x 1.3dia. cm (24 x 0.5")
The converter clamp supports the ultrasonic stack (converter and horn) and is easily adjusted to properly position the horn in the sample. The stand features a black enameled cast-iron base with zinc-plated rod.
Ordering information: Support stand with Rod is supplied without clamps, which must be purchased separately. Clamp orders include clamp only.