EP Scientific Silanized Screw Thread Vials, Thermo Scientific
Supplier: Thermo Scientific
SAA-SV2-2 SAA-SV4-2 SCA-SV4-2
89026-610CS 546.07 CAD
89026-610 89026-616 89026-618
EP Scientific Silanized Screw Thread Vials, Thermo Scientific
Vials
These vials are mainly used to preserve the integrity of certain materials or extracts stored in glass containers.
  • Trace organic analysis
  • Storage of materials prone to adhering to glass
  • Extraction glassware