Water, special quality for TOC

Supplier: EP SCIENTIFIC PRODUCTS

Synonyms: Hydrogen oxide

111-04A/C-TOC 112-01A/CTOC
EP111-04ACTOCCS 504.64 CAD
EP111-04ACTOC EP112-01A-CTOC
Water, special quality for TOC
Water

Manufactured, packaged and tested to exacting standards, these specialty waters are highly purified low TOC water in 1 L bottles or 4 L glass jugs.


  • Packaged in pre-cleaned amber glass containers inside Class 10 environment
  • Purity exceeds 17 MegaOhm resistivity
  • Low organic and inorganic impurities at time of packaging
  • Processing includes reverse osmosis, activated carbon, and ultraviolet TOC reduction
  • Filtered to 0.1 µm through hydrophobic membrane filters


Processing includes reverse osmosis, activated carbon, and ultraviolet TOC reduction.


Certifications: Certified to contain less-than 50 ppb TOC

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0 °C
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR