Xylene (mixture of isomers), Low_water 98,5-99,41 isomers plus ethylbenzene ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Dimethylbenzene

BDH1170-4LG BDH1176-204L
BDH1170-4LGCS 1644.09 CAD
BDH1170-4LG BDH1176-204L
Xylene (mixture of isomers), Low_water 98,5-99,41 isomers plus ethylbenzene ACS, VWR Chemicals BDH®
Xylene (mixture of isomers)

Suitable for general laboratory use, Histology and low-water applications.


  • Meets ACS Specifications.
  • Low water 0. 02% maximum.


The 204L steel drum has 2 and 3/4inch NPT bungs.

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Purity (Xylenes + Ethylbenzene 98.5% Min. 99.41 %
Ethylbenzene (by GC) Less than 25.0%
Color (APHA) Less than 10
Residue Less than 0.002%
Sub. Darkened by H2SO4 Passes Test
Sulfur Compounds (as S Less than 0.003%
Water (by K.F.) Less than 0.02%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR