StableTemp™ Silicone Rubber Heating Tapes, Cole-Parmer®
Supplier: Antyila Scientific
03111-12 03110-40 03110-84
97010-400EA 568.15 CAD
97010-400 97010-408 97010-424 97010-432 97010-440 97010-192 97010-416
StableTemp™ Silicone Rubber Heating Tapes, Cole-Parmer®
Tapes
Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in high-strength, hightemperature-resistant silicone rubber.
  • Moisture and chemical resistant
  • Maximum exposure temperature of 450 °F (232 °C)
  • Nongrounded
The 120 VAC tapes have a same-end lead configuration and are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug. Silicone rubber-insulated lead wires extend two feet from opposite ends of the tape to simplify connection to power source.