Microflex® 93-243 Gloves, Ansell
Supplier: Ansell Healthcare
870997 870998 870999 871000 871001 871002
75795-894BG 94.85 CAD
75795-894 75795-896 75795-898 75795-900 75795-984 75795-986
Microflex® 93-243 Gloves, Ansell
Gloves
Microflex® 93-243 Series Gloves are designed with an elbow-length cuff to provide additional protection to the upper arm and extra security against cross contamination, while preventing the glove from rolling down.
  • Ambidextrous
  • Textured grip
  • Powder-free
  • For single use only
Nitrile formulation offers greater puncture resistance and chemical resistance than natural rubber latex.
Microflex® 93-243 Series Gloves are well suited for use in a wide variety of industries, including automotive, chemical (basic, specialty, and petrochemical), electronics (More Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable Disposable
Latex/Latex-free Latex-free Latex-free
Powdered/Powder-free Powder-free Powder-free
Cuff style Beaded Beaded
Finger thickness 4.3 mil
Palm thickness 2.8 mil