Cole-Parmer® BriskHeat High-Temperature Heating Tapes, Antylia Scientific
Supplier: Antyila Scientific
36119-31 36119-30 36119-33
97010-316EA 156.97 CAD
97010-316 97010-312 97010-324 97010-332
Cole-Parmer® BriskHeat High-Temperature Heating Tapes, Antylia Scientific
Tapes
These high-temperature heating tapes are made of resistance wires that are double insulated with braided SAMOX yarns, then knit into flat tapes
  • Flexible
  • Maximum exposure temperature of 1400 °F (760 °C)
  • Standard-duty insulated design
  • Nongrounded
  • Direct-contact heaters for use on non-conductive surfaces such as glassware
Tapes include ties at each end, and a standard two-prong split plug.