Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific
Supplier: Antyila Scientific
36225-00 36225-02 36225-04 36225-06 36225-10 36225-30 36225-40 36225-50 36225-60 36225-70 36226-10 36226-20 36226-30
97010-916EA 843.19 CAD
97010-916 97010-920 97010-924 97010-928 97010-932 97010-936 97010-940 97010-944 97010-948 97010-952 97010-956 97010-960 97010-964 97010-968 97010-972 97010-976 97010-980 97010-984 97010-988 97010-992
Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific
Heaters Heating Mantles
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes.
  • Maximum temperature of 842°F (450°C)
  • Require a controlled electrical power source to prevent overheatin
  • Models for 120 VAC operation are CSA-certified
Hemispherical Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel supports (sold separately) are required to support mantle and allow attachment to ring stands. Supports have fabric strap basket to hold mantles;do not place mantle directly on ring stand.