Orion™ Total Alkalinity Test Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
700011-WA 700010 700012
23198-342CS 451.24 CAD
23198-342 34182-060 34182-064
Orion™ Total Alkalinity Test Kit, Thermo Scientific
Environmental Test Kits
Save time by eliminating additional equipment set-up, titrations, and calculations
  • Direct reading of total alkalinity from 0 to 200ppm as CaCO3, from the enclosed pH conversion chart
  • No interference from color or turbidity
The Orion™ Total Alkalinity Test Kit provides two-step direct measurement of total alkalinity and pH, using a pH electrode.
Ordering information: Test kits include one 475mL bottle of alkalinity reagent, one 475mL bottle of alkalinity standard/control, total alkalinity conversion chart, and application procedure.