Orion™ Lead Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9682BNWP 9482SC 9682BNWP
CA10010-824EA 2212 CAD
CA10010-824 10010-824 34117-936
Orion™ Lead Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure lead ions in aqueous solutions quickly, simply, accurately, and economically with Orion™ Lead Electrodes.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode ORIO9682BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.