Reactive Dye Conjugates, Biotium
Supplier: Biotium
90073 90103 90104 90105 90106 90107 90108 90109 90110 90115 90116 92000 92001
89139-650EA 420.98 CAD
89139-650 89139-706 89139-708 89139-710 89139-712 89139-714 89139-716 89139-718 89139-720 89139-730 89139-732 89139-794 89139-796
Reactive Dye Conjugates, Biotium
Protein Linking and Labeling Systems
Reactive dyes containing an amine functional group.
  • Aminooxy conjugates
  • Cadaverine conjugates
  • Can be used to label abasic sites in damaged DNA
Our aminooxy reagents such as aminooxy-5(6)-FAM react with aldehydes or ketones to form a stable hydrazone compound under mild conditions
Dye conjugates to PEO3-amine have a long water-soluble PEG spacer that separates the fluorescent dye and the amine functional group. These dyes can be coupled to activated carboxy groups or sulfonyl chlorides.
Cadaverine conjugates can be coupled to activated carboxy groups or sulfonyl chlorides.
Similar to ARP, aminooxy-5(6)-FAM can be used to label abasic sites in damaged DNA.