One-Step Lumitein™ Protein Gel Stain, Biotium
Supplier: Biotium
One-Step Lumitein™
21005-1L 21004-4L 21005-4L 21004-1L
10809-756EA 931.65 CAD
10809-756 10809-754 75832-774 75832-776
One-Step Lumitein™ Protein Gel Stain, Biotium
Stains Electrophoresis Stains
One-Step Lumitein™ Protein Gel Stain is a ready-to-use luminescent red protein gel stain. It is a dramatically improved version of our original Lumitein™ protein gel stain in terms of convenience and safety. One-Step Lumitein™ can detect as little as approximately 1-10 ng of protein per band.
  • One-step staining procedure, no fixation required
  • Non-toxic formulation is safe for easy disposal
  • Sensitive protein detection down to 1-10 ng
  • Image with a UV transilluminator, a blue light transilluminator, or laser-based gel scanner
  • Store at 4°C
Fast and simple procedure
One-Step Lumitein™ gel staining requires only a single 5-60 minute staining step without fixation. Destaining is optional. One-Step Lumitein™ can detect as little as approximately 1-10 ng of protein per band depending on the staining method used, although staining intensity varies between proteins. Staining is fully compatible with mass spectrometry and Edman-based sequencing. Staining can be imaged using a UV transilluminator with EtBr or SYBR filter set, a blue light transilluminator, or a laser-based gel scanner.
One-Step Lumitein™ is non-toxic
One-Step Lumitein™ Protein Gel Stain offers safer handling and ease of disposal compared to other protein gel stains, because it is an aqueous-based solution that does not contain hazardous methanol or acetic acid. One-Step stains are certified under CCR Title 22 as non-toxic to the environment for drain disposal after a simple pH neutralization step. Furthermore, One-Step Lumitein™ costs significantly less than other fluorescent protein gel stains. One-Step Lumitein™ Protein Gel Stain is available in 1 liter bottle or 4 liter Cubitainer® sizes.
One-Step Lumitein™: Replacement for SYPRO® Ruby Protein Gel Stain
One-Step Lumitein™ UV: Replacement for Oriole™ and Flamingo™ Protein Gel Stains
Also see our One-Step Blue™ Protein Gel Stain, a rapid, easy-to-use, non-toxic alternative to Coomassie staining for visible blue protein staining and optional near-infrared fluorescence-based gel imaging.