Mix-n-Stain™ CF® Dye Small Ligand Labeling Kits, Biotium
Supplier: Biotium
Easily label SNAP-tag®, CLIP-tag™, HaloTag®, or other ligands with fluorescent dyes, no purification needed. Cell membrane-permeant and impermeant dye options for intracellular or surface targets.
- For SNAP-tag®, CLIP-tag™ and HaloTag®, or other ligands with an aliphatic amine
- Label 100 nmol ligand
- Reaction takes ~30 minutes with minimal hands-on time and no purification
- Choose from 15 bright and photostable CF® Dyes
- Cell membrane-permeant or impermeant options for intracellular or surface targets
Mix-n-Stain™ small ligand labeling kits are designed for rapid labeling of low molecular weight (MW ~150 to 5000), high affinity biological ligands or substrates.
Biotium also offers TMP-PEG3-Amine for making fluorescent TMP ligands. Also see Biotium's Mix-n-Stain™ antibody labeling kits available with bright and photostable CF® Dyes and other labels.
SNAP-tag and CLIP-tag are trademarks or registered trademarks of New England BioLabs; HaloTag is a registered trademark of Promega Corporation.