Cimerac™ Basic Stirrers, Thermo Scientific
Supplier: Thermo Fisher Scientific
S194615 S194925
97027-348EA 528.1 CAD
97027-348 97027-354
Cimerac™ Basic Stirrers, Thermo Scientific
Stirrers Magnetic Stirrers
Reach stirring speeds from 250 to 2500 rpm with Cimarec™ Basic Stirrers
  • Control knobs allow precise speed setting
  • Top-plate design reduces chance of breakage
  • Spillaway design directs spills away from internal components
  • Integral ring-stand holder accommodates 1.3 cm (1/2") diameter support rod
Ideal for economical, general-purpose use.
Delivery information: Stirrers include TFE-coated stir bar.