Dimethyl sulfoxide, anhydrous ≥99.9%

Supplier: Biotium

Synonyms: Methyl sulfoxideDMSODimethyl(oxido)sulfurMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphurMethanesulfinylmethane

90082
89139-666EA 68.49 CAD
89139-666
Dimethyl sulfoxide, anhydrous ≥99.9%
Dimethyl sulphoxide

Anhydrous DMSO (dimethyl sulfoxide; methyl sulfoxide) is recommended for preparing stock solutions.


  • Prepare stock solutions of AM ester dyes
  • Stock solutions of reactive dyes and other related, moisture-sensitive products


Anhydrous DMSO (dimethyl sulfoxide; methyl sulfoxide) is recommended for preparing stock solutions of AM ester dyes, reactive dyes and other related moisture-sensitive products.

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
https://ca.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg

Order Now

Specification Test Results

Color & Form Colorless liquid
Purity ≥ 99.9%
Water Max 0.005%
Residual on evaporation Max 0.0005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR