Reactive Oxygen Species Indicators, Biotium
Supplier: Biotium
10055 10056 10057 10058 10056-1
89138-254EA 265.17 CAD
89138-254 89138-256 89138-258 89138-260 71004-310
Reactive Oxygen Species Indicators, Biotium
Protein Linking and Labeling Systems
Commonly used fluorescent dyes for detection of oxidative activities in cells.
  • Detect reactive oxygen species including superoxide
  • Detect oxidative activities in viable cells, including respiratory burst in phagocyte
Dihydrorhodamine 123 is the reduced form of rhodamine 123, which is a commonly used fluorescent mitochondrial dye
Dihydrorhodamine 123 dihydrochloride is functionally equivalent to dihydrorhodamine 123 but with increased stability toward air oxidation and light during storage.
Dihydroethidium (also called hydroethidium) is the chemically reduced form of the commonly used DNA dye ethidium bromide. The probe is useful to detect oxidative activities in viable cells, including respiratory burst in phagocytes. Dihydroethidium itself has blue fluorescence (λ Ex/λ Em: 355/420nm) in cells, while the oxidized form ethidium has red fluorescence (λ Ex/λ Em: 518/605 nm) upon DNA intercalation.
H2DCFDA (2′,7′-Dichlorodihydrofluorescein diacetate) is a useful fluorogenic reagent to detect reactive oxygen intermediates in cells. On oxidation, H2DCFDA becomes the highly green fluorescent 2′,7′-dichlorofluorescein. λEx/λ Em (pH>6) = 504/529 nm (end product)
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more