Sodium sulfate, anhydrous ≥99.0% USP, Ph. Eur. endotoxin tested

Supplier: Avantor Performance Materials

VWRB836-12KG VWRB836-500G
VWRB836-12KGEA 1535.73 CAD
VWRB836-12KG VWRB836-2.5KG VWRB836-500G
Sodium sulfate, anhydrous ≥99.0% USP, Ph. Eur. endotoxin tested
Sodium sulphate

Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available


Material is of non-animal, non-human origin

Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
MDL Number: MFCD00003504
CAS Number: 7757-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity or Alkalinity (USP) PASS
Assay (USP) ≥99.0%
Endotoxins (USP) ≤2.5 EU/g
Chloride (USP) 221 ≤0.02%
Heavy Metals (USP) 231 ≤0.001%
Identification (USP) PASS
Loss on Drying (USP) 731 ≤0.5%
Acidity or Alkalinity (EP) PASS
Appearance of Solution (EP) PASS
Assay (EP) 98.5 – 101.0%
Calcium (EP) (243) ≤ 450ppm
Chlorides (EP) (244) ≤ 450ppm
Heavy Metals (EP) (248) ≤ 45ppm
Identification A (EP) PASS
Identification B (EP) PASS
Identification C (EP) PASS
Iron (EP) (249) ≤ 90ppm
Loss on Drying (EP) (2232) ≤ 0.5%
Magnesium (EP) ≤ 200ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR