BRANDplates® Insert System for Cell Culture, BrandTech®
Supplier: Brandtech
782840 782841 782800 782801 782810 782811 782820 782821 782870 782871 782880 782881 782890 782891 782860 782861
10215-418PK 643.07 CAD
10215-418 10215-420 10215-434 10215-436 10215-442 10215-444 10215-450 10215-452 10215-466 10215-468 10215-474 10215-476 10215-478 10215-480 10215-482 10215-484
BRANDplates® Insert System for Cell Culture, BrandTech®
Microplates Insert Plates and Inserts
BRANDplates® Insert System includes a choice of two types of cell culture plates and two types of cell culture inserts for co-culture, migration/invasion assays, chemotaxis assays, and 3D tissue preparation.
  • Inserts with matched plates
  • 24-well design for individual assays
  • Special 6-well design allows exact duplication of external conditions
  • Available with or without inlet openings
Plates are available in 24-well or specialized 6-well formats as non-treated sterile (pureGrade™), or with a cellGrade™ plus surface for enhanced cell culture. TheMore Product Information