EP Scientific, Depyrogenated Jars, with Closure, Thermo Scientific
Supplier: Thermo Scientific
117-2L/PF 132-16C/PF 117-4L/PF 130-02C/PF 130-04C/PF C20-02A/PF C20-04A/PF
89026-602CS 929.96 CAD
89026-602 89026-608 EP117-4L-PF EP130-02C-PF EP130-04C-PF EPC20-02A-PF EPC20-04A-PF
EP Scientific, Depyrogenated Jars, with Closure, Thermo Scientific
Jars General Use Jars
Thermo Scientific™ Depyrogenated Glass Containers are specially prepared to meet endotoxin levels of less than 0.06 EU/mL for use in terminal sterilization, water sampling and product storage.
  • Used for water sampling
  • Cleaning and packaging is performed in a Class 100/10 Cleanroom
  • Packaging and storing laboratory reagents, samples and medias
Recommended for endotoxin-free applications. Thermo Scientific also offers glass vials specially prepared to meet endotoxin levels of less than 0.06 EU/mL.
Certifications: Certificate of Analysis is includedMore Product Information